missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium iodide, 98%, Thermo Scientific™

Product Code. 10055363 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10055363 500 g Glass bottle 500g
10002850 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10055363 Supplier Thermo Scientific Acros Supplier No. 216675000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68-05-3
Molecular Formula C8H20IN
Molecular Weight (g/mol) 257.16
MDL Number MFCD00011829
InChI Key UQFSVBXCNGCBBW-UHFFFAOYSA-M
Synonym tetraethylammonium iodide, tetamon iodide, tetramon j, ethanaminium, n,n,n-triethyl-, iodide, n,n,n-triethylethanaminium iodide, ammonium, tetraethyl-, iodide, teai, tetraethylammoniumiodide, tetraethylazanium iodide, tetraethyl ammonium iodide
PubChem CID 6225
IUPAC Name tetraethylazanium iodide
SMILES [I-].CC[N+](CC)(CC)CC

Specifications

Color White to Cream
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (C2H5)4NI
Quantity 500 g
Beilstein 04, 104
Fieser 12,465; 13,46
Merck Index 10, 9027
Formula Weight 257.15
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material Tetraethylammonium iodide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.