missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium nitrate, 98%

Product Code. 10055203 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
Packaging:
Glass bottle
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10055203 10 g Glass bottle 10g
10381161 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10055203 Supplier Thermo Scientific Acros Supplier No. 420310100
  • ONLINE EXCLUSIVE PRICE
    70.60 €
    68.50 € / 10g
    Save 2.10 €
    2% Off
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 1941-26-0 | C8H20N2O3 | 192.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1941-26-0
Molecular Formula C8H20N2O3
Molecular Weight (g/mol) 192.26
MDL Number MFCD00041978
InChI Key JTJKNAJRGLQKDZ-UHFFFAOYSA-N
Synonym tetraethylammonium nitrate, ethanaminium, n,n,n-triethyl-, nitrate, ammonium, tetraethyl-, nitrate, ethanaminium, n,n,n-triethyl-, nitrate 1:1, tetraethylazanium nitrate, tetraethyl ammonium nitrate, tetraethylammonium nitrate w/w aqueous solution
PubChem CID 74744
IUPAC Name tetraethylazanium;nitrate
SMILES [O-][N+]([O-])=O.CC[N+](CC)(CC)CC

Specifications

Melting Point >240.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (C2H5)4NNO3
Quantity 10 g
Solubility Information Solubility in water: >50% (20°C).
Formula Weight 192.26
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Tetraethylammonium nitrate, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Tetraethylammonium nitrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.