missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium Tetrafluoroborate, TRC

Product Code. 30409943
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30409943 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30409943 Supplier Toronto Research Chemicals Supplier No. TRCT29229525G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetraethylammonium Tetrafluoroborate
CAS 429-06-1
Molecular Formula C8 H20 B F4 N3
Quantity 25 g
Synonym Digirena, LES 100, Lipaste AN/EAF 1, Lipaste P/EAFIN, Merck Supercapacitor Electrolyte LES 100, NSC 231537, N,N,N-Triethylethanaminium Tetrafluoroborate
InChI Formula InChI=1S/C8H20N.BF4/c1-5-9(6-2,7-3)8-4;2-1(3,4)5/h5-8H2,1-4H3;/q+1;-1
SMILES CC[N+](CC)(CC)CC.F[B-](F)(F)F
IUPAC Name tetraethylazanium;tetrafluoroborate
Molecular Weight (g/mol) 217.06
Formula Weight 217.1625
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.