missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylene glycol dimethyl ether, 99%

Product Code. 10439470 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
Unit Size:
1kg
2.5kg
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10439470 1 kg Glass bottle 1kg
10124000 250 g Glass bottle 250g
11904741 2.5 kg Glass bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10439470 Supplier Thermo Scientific Acros Supplier No. 174110010
  • ONLINE EXCLUSIVE PRICE
    133.00 €
    122.00 € / 1kg
    Save 11.00 €
    8% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 143-24-8 | C10H22O5 | 222.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143-24-8
Molecular Formula C10H22O5
Molecular Weight (g/mol) 222.28
MDL Number MFCD00008505
InChI Key ZUHZGEOKBKGPSW-UHFFFAOYSA-N
Synonym tetraglyme, tetraethylene glycol dimethyl ether, 2,5,8,11,14-pentaoxapentadecane, dimethoxytetraglycol, glyme 5, ansul ether 181at, dimethoxytetraethylene glycol, nissan uniox mm 200, bis 2-2-methoxyethoxy ethyl ether, methyltetraglyme200
PubChem CID 8925
ChEBI CHEBI:46785
IUPAC Name 1-methoxy-2-[2-[2-(2-methoxyethoxy)ethoxy]ethoxy]ethane
SMILES COCCOCCOCCOCCOC

Specifications

Acidity 0.015% max.
Melting Point -30.0°C
Density 1.0090g/mL
Boiling Point 275.0°C to 276.0°C
Flash Point 141°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(OCH2CH2)4OCH3
Refractive Index 1.4310 to 1.4330
Quantity 1 kg
Beilstein 01,III,2107
Merck Index 15,9353
Specific Gravity 1.009
Solubility Information Solubility in water: soluble. Other solubilities: miscible with hydrocarbon solvents
Viscosity 4.1 mPa.s (20°C)
Formula Weight 222.28
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Tetraethylene glycol dimethyl ether
Show More Show Less
compliance-icons
Product Identifier
  • Tetraethylene glycol dimethyl ether
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.