missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Tetraethylene Glycol Monooctyl Ether (C8E4), LC/MS grade

Product Code. 30245088 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30245088 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30245088 Supplier Thermo Scientific™ Supplier No. A65524

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Tetraethylene Glycol Monooctyl Ether (C8E4) is a non-ionic surfactant that possesses unique properties making it suitable for solubilization of sensitive biomolecules without significant denaturation or disruption of their structure and function.

Tetraethylene Glycol Monooctyl Ether (C8E4) is a non-ionic surfactant that possesses unique properties making it suitable for solubilization of sensitive biomolecules without significant denaturation or disruption of their structure and function. It has good solubility in both aqueous and organic solvents, allowing for versatile applications. C8E4 can be used as a detergent in native MS to solubilize and stabilize biomolecules, particularly membrane proteins or protein complexes, in aqueous solutions. It helps maintain the native conformation of these biomolecules and facilitates their transfer into the gas phase during the MS analysis.

Features of C8E4 include:
• Non-ionic detergent
• Soluble in both water and organic solvents
• Enhanced stability of solubilized biomolecules, including membrane proteins
• High-purity compound
• Native mass spec compatible

Applications
C8E4 is versatile and can be used in various applications in addition to native MS, including protein purification, crystallization, and structural biology studies. It is compatible with a wide range of biomolecules and can be employed for both soluble and membrane protein research.

Usage
Optimizing the concentration of C8E4 is crucial to ensure effective solubilization and stabilization of the biomolecules while minimizing detergent interference in the mass spectrometry analysis. Additionally, the specific conditions and parameters of the native MS experiment, such as the choice of ionization source and mass spectrometer settings, should be carefully considered and optimized for each specific application.

Properties
CMC: (0.1% NaCl) ~8 mM (0.25%)
Aggregation number: (H2O) ~82
Purity: ≥99% by HPLC analysis
pH: 5–8 (5% solution in water)

TRUSTED_SUSTAINABILITY

Specifications

Color Clear
CAS 19327-39-0
Molecular Formula C16H34O5
Content And Storage Store <-20°C.
Form Liquid
For Use With (Application) LC-MS
Quantity 10 mL
Molecular Weight (g/mol) 306.5
pH 5 to 8 (5% solution in water)
Purity ≥99%

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.