missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylene glycol monooctyl ether

Product Code. 15444879
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15444879 1 g 1g
15454879 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15444879 Supplier Thermo Scientific Alfa Aesar Supplier No. J62496.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Reagent for solubilizing membrane proteins | CAS: 19327-39-0 | C16H34O5 | 306.443 g/mol

Reagent for solubilizing membrane proteins

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19327-39-0
Molecular Formula C16H34O5
Molecular Weight (g/mol) 306.443
MDL Number MFCD00043033
InChI Key FEOZZFHAVXYAMB-UHFFFAOYSA-N
Synonym 3,6,9,12-tetraoxaeicosan-1-ol, 3,6,9,12-tetraoxaicosan-1-ol, tetraethylene glycol monooctyl ether, hydroxyethyloxy tri ethyloxy octane, tetraethylene glycol monoctyl ether, c8e, octyltetraglycol, octyltetraethyleneoxide, d06pvf, octyl tetraethylene glycol ether
PubChem CID 5414
ChEBI CHEBI:41323
IUPAC Name 2-[2-[2-(2-octoxyethoxy)ethoxy]ethoxy]ethanol
SMILES CCCCCCCCOCCOCCOCCOCCO

Specifications

Color Colorless
Density 0.969
Linear Formula CH3(CH2)7(OCH2CH2)4OH
Quantity 1 g
Beilstein 1781226
Formula Weight 306.44
Physical Form Liquid
Chemical Name or Material Tetraethylene glycol monooctyl ether
compliance-icons
Product Identifier
  • Tetraethylene glycol monooctyl ether
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 3,6,9,12-Tetraoxaeicosan-1-ol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.