missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylene Glycol, TRC

Product Code. 30330664
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30330664 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30330664 Supplier Toronto Research Chemicals Supplier No. TRCB44382010G
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetraethylene glycol
CAS 112-60-7
Molecular Formula C8 H18 O5
Quantity 10 g
Synonym Tetraethylene glycol (6CI,7CI,8CI), 2,2'-[Oxybis(2,1-ethanediyloxy)]bis[ethanol], 1,11-Dihydroxy-3,6,9-trioxaundecane, 2,2'-((Oxybis(ethane-2,1-diyl))bis(oxy))diethanol, 2-[2-[2-(2-Hydroxyethoxy)ethoxy]ethoxy]ethan-1-ol, 2-[2-[2-(2-Hydroxyethoxy)ethoxy]ethoxy]ethanol, 3,6,9-Trioxaundecane-1,11-diol, Bis[2-(2-hydroxyethoxy)ethyl] ether, Hi-Dry, NSC 1262, TEG
InChI Formula InChI=1S/C8H18O5/c9-1-3-11-5-7-13-8-6-12-4-2-10/h9-10H,1-8H2
SMILES OCCOCCOCCOCCO
IUPAC Name 2-[2-[2-(2-hydroxyethoxy)ethoxy]ethoxy]ethanol
Molecular Weight (g/mol) 194.23
Formula Weight 194.1154
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.