missing translation for 'onlineSavingsMsg'
Learn More

Tetrafluorosuccinic acid, 97%

Product Code. 11474437
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11474437

Brand: Thermo Scientific Alfa Aesar B20462.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 377-38-8 | C4H2F4O4 | 190.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 377-38-8
Molecular Formula C4H2F4O4
Molecular Weight (g/mol) 190.05
MDL Number MFCD00042430
InChI Key YUDUFRYTKFGQCL-UHFFFAOYSA-N
PubChem CID 67833
IUPAC Name 2,2,3,3-tetrafluorobutanedioic acid
SMILES C(=O)(C(C(C(=O)O)(F)F)(F)F)O

Specifications

Melting Point ∼115°C
Density 1.4
Boiling Point 150°C (15 mmHg)
Linear Formula HO2CCF2CF2CO2H
Quantity 5 g
UN Number UN3261
Beilstein 1790197
Synonym tetrafluorosuccinic acid, perfluorosuccinic acid, butanedioic acid, tetrafluoro, succinic acid, perfluoro, succinic acid, tetrafluoro, tetrafluorobutanedioic acid, butanedioic acid, 2,2,3,3-tetrafluoro, 2,2,3,3-tetrafluorosuccinic acid, butanedioic acid,2,2,3,3-tetrafluoro, perfluorosuccinate
Formula Weight 190.05
Percent Purity 97%
Chemical Name or Material Tetrafluorosuccinic acid
Show More Show Less
compliance-icons
Product Identifier
  • Tetrafluorosuccinic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Tetrafluorosuccinicacid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.