missing translation for 'onlineSavingsMsg'
Learn More

Tetraglyme, TRC

Product Code. 30135143
Change view
Click to view available options
Quantity:
100 mg
1 g
10 g
Unit Size:
100mg
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135143 10 g 10g
18091220 100 mg 100mg
18022420 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135143 Supplier Toronto Research Chemicals Supplier No. TRCT29299510G
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143-24-8
Molecular Formula C10 H22 O5
Molecular Weight (g/mol) 222.28
Synonym Dimethoxytetraethylene Glycol, 2,5,8,11,14-Pentaoxapentadecane, 1-Methoxy-2-[2-[2-(2-methoxyethoxy)ethoxy]ethoxy]ethane, Ansul Ether 181AT, Bis[2-(2-methoxyethoxy)ethyl] ether, Dimethoxytetraethylene glycol, Dimethoxytetraglycol, E 181, E 181 (ether), Ether, bis[2-(2-methoxyethoxy)ethyl], Glyme 5, Hisolve MTEM, Methyltetraglyme, NSC 65624, Nissan Uniox MM 200, TEGDME, Tetraethylene glycol dimethyl ether, Tetraglyme, tetraethylene glycol di-Me ether
IUPAC Name 1-methoxy-2-[2-[2-(2-methoxyethoxy)ethoxy]ethoxy]ethane
SMILES COCCOCCOCCOCCOC

Specifications

Quantity 10 g
InChI Formula InChI=1S/C10H22O5/c1-11-3-5-13-7-9-15-10-8-14-6-4-12-2/h3-10H2,1-2H3
Formula Weight 222.1467
Chemical Name or Material Tetraglyme
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.