missing translation for 'onlineSavingsMsg'
Learn More

Tetraheptylammonium Bromide, TRC

Product Code. 30134389
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134389 5 g 5g
30011028 10 g 10g
30005459 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134389 Supplier Toronto Research Chemicals Supplier No. TRCT2942855G
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4368-51-8
Molecular Formula C28H60BrN
Molecular Weight (g/mol) 490.69
Synonym N,N,N-Triheptyl-1-heptanaminium Bromide;Tetraheptylammonium Bromide, Tetraheptylammonium Bromide;Tetra(n-heptyl)ammonium Bromide
IUPAC Name tetraheptylazanium;bromide
SMILES [Br-].CCCCCCC[N+](CCCCCCC)(CCCCCCC)CCCCCCC

Specifications

Quantity 5 g
InChI Formula InChI=1S/C28H60N.BrH/c1-5-9-13-17-21-25-29(26-22-18-14-10-6-2,27-23-19-15-11-7-3)28-24-20-16-12-8-4;/h5-28H2,1-4H3;1H/q+1;/p-1
Formula Weight 489.3909
Chemical Name or Material Tetraheptylammonium Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.