missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydro-2H-pyran-2-ol, 90%, cyclized form of 5-Hydroxypentanal

Product Code. 10381253 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10381253 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10381253 Supplier Thermo Scientific Acros Supplier No. 387900250
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 694-54-2 | C5H10O2 | 102.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 694-54-2
Color Colorless
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Molecular Formula C5H10O2
MDL Number MFCD02683093
Quantity 25 g
Synonym 2-hydroxytetrahydropyran, tetrahydro-2h-pyran-2-ol, oxanol, 2h-pyran-2-ol, tetrahydro, 2-tetrahydropyranol, tetrahydropyran-2-ol, tetrahydro-2-hydroxy-2h-pyran, tetrahydro-2-pyranol, tetrahydropyranyl alcohol, 2h-pyran-2-ol,tetrahydro
InChI Key CELWCAITJAEQNL-UHFFFAOYSA-N
SMILES C1CCOC(C1)O
IUPAC Name oxan-2-ol
Molecular Weight (g/mol) 102.13
PubChem CID 136505
Formula Weight 102.13
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material Tetrahydro-2H-pyran-2-ol
Show More Show Less
compliance-icons
Product Identifier
  • Tetrahydro-2H-pyran-2-ol, cyclized form of 5-Hydroxypentanal
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.