missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydro-2H-thiopyran-4-ol, 97%, Thermo Scientific™

Product Code. 10295714
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10295714 250 mg 250mg
10412854 1 g 1g
10785764 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10295714 Supplier Thermo Scientific Maybridge Supplier No. CC61915CB
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 29683-23-6
Molecular Formula C5H10OS
Molecular Weight (g/mol) 118.19
MDL Number MFCD02942489
InChI Key YODQQARABJQLIP-UHFFFAOYSA-N
Synonym tetrahydro-2h-thiopyran-4-ol, tetrahydrothiopyran-4-ol, thiacyclohexane-4-ol, 2h-thiopyran-4-ol, tetrahydro, 4-hydroxythiane, acmc-209ydy, tetrahydrothiapyran-4-ol, 4-hydroxytetrahydrothiopyran, 4-hydroxy-tetrahydrothiopyran, tetrahydro-4h-thiopyran-4-ol
PubChem CID 576296
IUPAC Name thian-4-ol
SMILES OC1CCSCC1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 250 mg
Formula Weight 118.2
Percent Purity 97%
Chemical Name or Material Tetrahydro-2H-thiopyran-4-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.