missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydro-3-furoic Acid, TRC

Product Code. 30415423
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30415423 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30415423 Supplier Toronto Research Chemicals Supplier No. TRCT2936105G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetrahydro-3-furoic Acid
CAS 89364-31-8
Molecular Formula C5 H8 O3
Quantity 5 g
Synonym 3-Furancarboxylic acid','tetrahydro-, 3-Furoic acid','tetrahydro- (6CI,7CI), Tetrahydro-3-furancarboxylic acid, 1,2,3,4-Tetrahydrofuran-3-carboxylic acid, Tetrahydro-3-furanoic acid, Tetrahydro-3-furoic acid, Tetrahydrofuran-4-carboxylic acid
InChI Formula InChI=1S/C5H8O3/c6-5(7)4-1-2-8-3-4/h4H,1-3H2,(H,6,7)
SMILES OC(=O)C1CCOC1
IUPAC Name oxolane-3-carboxylic acid
Molecular Weight (g/mol) 116.12
Formula Weight 116.0473
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.