missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydro-4H-thiopyran-4-one, 99%

Product Code. 11472578
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11472578 1 g 1g
11482578 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11472578 Supplier Thermo Scientific Alfa Aesar Supplier No. A15872.03
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 1072-72-6 | C5H8OS | 116.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1072-72-6
Molecular Formula C5H8OS
Molecular Weight (g/mol) 116.18
MDL Number MFCD00006660
InChI Key OVRJVKCZJCNSOW-UHFFFAOYSA-N
Synonym tetrahydrothiopyran-4-one, tetrahydro-4h-thiopyran-4-one, 4-oxothiane, 4-thiacyclohexanone, penthianone, 4h-thiopyran-4-one, tetrahydro, 1-thiacyclohexan-4-one, dihydro-2h-thiopyran-4 3h-one, 4-oxotetrahydrothiopyran, tetrahydro-4-thiopyrone
PubChem CID 66173
IUPAC Name thian-4-one
SMILES O=C1CCSCC1

Specifications

Melting Point 60°C to 64°C
Odor Weak
Quantity 1 g
Formula Weight 116.18
Percent Purity 99%
Chemical Name or Material Tetrahydro-4H-thiopyran-4-one

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.