missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydro-beta-carboline, 98%

Product Code. 10225164 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10225164 1 g Glass bottle 1g
10584954 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10225164 Supplier Thermo Scientific Acros Supplier No. 427580010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy

CAS: 16502-01-5 | C11H13ClN2 | 208.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16502-01-5
Molecular Formula C11H13ClN2
Molecular Weight (g/mol) 208.69
MDL Number MFCD00004954
InChI Key PHLJRXUBLWEPCM-UHFFFAOYSA-N
Synonym tryptoline, 2,3,4,9-tetrahydro-1h-pyrido 3,4-b indole, noreleagnine, 1,2,3,4-tetrahydro-9h-pyrido 3,4-b indole, 1,2,3,4-tetrahydro-beta-carboline, tetrahydro-beta-carboline, thbc, tetrahydronorharman, unii-65027tmi0h, 1h,2h,3h,4h,9h-pyrido 3,4-b indole
PubChem CID 107838
IUPAC Name 2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole
SMILES [H+].[Cl-].C1CC2=C(CN1)NC1=CC=CC=C21

Specifications

Melting Point 206°C to 208°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 172.23
Percent Purity 98%
Physical Form Crystals or Powder
Chemical Name or Material Tetrahydro-β-carboline
compliance-icons
Product Identifier
  • Tetrahydro-beta-carboline
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.