missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydro Furosemide, TRC

Product Code. 30409174
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30409174 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30409174 Supplier Toronto Research Chemicals Supplier No. TRCT29354050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetrahydro Furosemide
CAS 4793-38-8
Purity Grade Notes HPLC
Molecular Formula C12 H15 Cl N2 O5 S
Quantity 50 mg
Synonym 5-(Aminosulfonyl)-4-chloro-2-[[(tetrahydro-2-furanyl)methyl]amino]benzoic acid, Anthranilic acid','4-chloro-5-sulfamoyl-N-(tetrahydrofurfuryl)- (7CI,8CI), Furosemide Imp. F (EP)
InChI Formula InChI=1S/C12H15ClN2O5S/c13-9-5-10(15-6-7-2-1-3-20-7)8(12(16)17)4-11(9)21(14,18)19/h4-5,7,15H,1-3,6H2,(H,16,17)(H2,14,18,19)
SMILES NS(=O)(=O)c1cc(C(=O)O)c(NCC2CCCO2)cc1Cl
IUPAC Name 4-chloro-2-(oxolan-2-ylmethylamino)-5-sulfamoylbenzoic acid
Molecular Weight (g/mol) 334.78
Formula Weight 334.039
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.