missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofuran,Solstice Biosyn™

Product Code. 16811250
Change view
Click to view available options
Packaging:
Glass Bottle
Quantity:
2.5 L
4 x 2.5 L
Unit Size:
1 L
10 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging Quantity Unit Size
16811250 Glass Bottle 2.5 L 1 L
16821250 Glass Bottle 4 x 2.5 L 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16811250 Supplier Burdick & Jackson BioSyn Supplier No. BB3442.5L
  • ONLINE EXCLUSIVE PRICE
    416.00 €
    375.78 € / 1 L
    Save 40.22 €
    9% Off
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For biosynthesis and applications that require low water content. Contains BHT, > 99.9% purity.

  • Ultra-low water specifications
  • Low amine specifications
  • Low particulate levels
  • High solvent purity
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Weight (g/mol) 72.11

Specifications

pH Neutral
Melting Point -108°C
Color Colorless
Density 0.89 g/cm3 (20°C)
Quantity 2.5 L
Explosion Limit 1.5% to 12% (V)
Flash Point -21°C
Packaging Glass Bottle
Physical Form Liquid
Product Line BioSyn
Vapor Pressure 200 hPa (20°C)
Viscosity 0.48 mPa.s (20°C)
UN Number UN2056
Boiling Point 65°C (1013 hPa)
Percent Purity >99.9%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.