missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofuran CHROMASOLV™ Plus, inhibitor-free, for HPLC, ≥99.9%, Solstice

Product Code. 15674600
Change view
Click to view available options
Quantity:
100 mL
1 L
2 L
2.5 L
4 x 2.5 L
6 x 1 L
Unit Size:
1 L
100mL
2 L
2.5 L
Pack of 4
Pack of 6
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15674600 1 L 1 L
15664600 100 mL 100mL
15694600 2 L 2 L
15684600 2.5 L 2.5 L
15614610 6 x 1 L Pack of 6
15604610 4 x 2.5 L Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
May form explosive peroxides.
Product Code. 15674600 Supplier Honeywell-Chromasolv Supplier No. 348651L
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
May form explosive peroxides.

Suitable for HPLC, spectrophotometry, environmental testing.

  • For HPLC, spectrophotometry, and environmental analysis
  • Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
  • Low UV impurity profile
  • Low impurity gradient LC baseline
  • Very low levels of non-volatile impurities
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-99-9
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00005356
InChI Key WYURNTSHIVDZCO-UHFFFAOYSA-N
Synonym tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
PubChem CID 8028
ChEBI CHEBI:26911
IUPAC Name oxolane
SMILES C1CCOC1

Specifications

Melting Point -108°C
Absorbance absorption/300nm ≤0.02
Density 0.889g/mL at 25°C
Quantity 1 L
Refractive Index n20/D 1.407
Linear Formula C4H8O
Vapor Pressure 143mmHg (20°C)
Beilstein 102391
Boiling Point 65°C to 67°C
Formula Weight 72.11g/mol
Percent Purity ≥99.9%
Grade CHROMASOLV™ Plus
Chemical Name or Material Tetrahydrofuran, inhibitor-free, for HPLC
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tetrahydrofuran
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H351-Suspected of causing cancer.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: tetrahydrofuran
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.