missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofuran-d{8}, 100%(Isotopic), Thermo Scientific Chemicals

Product Code. 11354448
Change view
Click to view available options
Quantity:
10 Ea.
2 Ea.
Unit Size:
Pack of 10
Pack of 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11354448 2 Ea. Pack of 2
11364448 10 Ea. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11354448 Supplier Thermo Scientific Alfa Aesar Supplier No. 042348.SA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Tetrahydrofuran-d_8 is a deuterated THF that is used in various synthesis reactions and also used as a precursor for various polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1693-74-9
Molecular Formula C4H8O
Molecular Weight (g/mol) 80.16
MDL Number MFCD00044238
InChI Key WYURNTSHIVDZCO-SVYQBANQSA-N
Synonym tetrahydrofuran-d8, deuterated thf, octadeuterotetrahydrofuran, deuterated tetrahydrofuran, tetrahydrofuran-d8 thf-d8, tetrahydrofuran-d8, ≥99.5 atom % d, 2h4 tetrahydro 2h4 furan, thf-d8, 2h8 tetrahydrofuran, furan-d4-, tetrahydro-d4
PubChem CID 80290
IUPAC Name 2,2,3,3,4,4,5,5-octadeuteriooxolane
SMILES [2H]C1([2H])OC([2H])([2H])C([2H])([2H])C1([2H])[2H]

Specifications

Physical Form Liquid
Color Colorless
Density 0.985g/mL
Flash Point −17°C (−1°F)
Packaging Prescored Ampule
Quantity 2 Ea.
Melting Point -106°C
Boiling Point 66°C
UN Number UN2056
Formula Weight 80.16
Percent Purity 100%
Chemical Name or Material Tetrahydrofuran-d8, Isotopic
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.