missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofuran-D8, for NMR, 99.5 atom % D

Product Code. 16368186 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
10 mL
25 mL
Packaging:
Glass Bottle
Unit Size:
10mL
1mL
25mL
5mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
16368186 25 mL Glass Bottle 25mL
10469660 1 mL Glass Bottle 1mL
10712611 5 mL Glass Bottle 5mL
10712801 10 mL Glass Bottle 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 16368186 Supplier Thermo Scientific Acros Supplier No. 180130250
  • ONLINE EXCLUSIVE PRICE
    970.00 €
    938.00 € / 25mL
    Save 32.00 €
    3% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 1693-74-9 | C4H8O | 80.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1693-74-9
Molecular Formula C4H8O
Molecular Weight (g/mol) 80.16
MDL Number MFCD00044238
InChI Key WYURNTSHIVDZCO-SVYQBANQSA-N
Synonym 2h8 tetrahydrofuran, tetrahydrofuran-d8, ≥99.5 atom % d, furan-d4-, tetrahydro-d4, tetrahydrofuran-d8, octadeuterotetrahydrofuran, deuterated tetrahydrofuran, tetrahydrofuran-d8 thf-d8, thf-d8, 2h4 tetrahydro 2h4 furan, deuterated thf
PubChem CID 80290
SMILES [2H]C1([2H])OC([2H])([2H])C([2H])([2H])C1([2H])[2H]

Specifications

Physical Form Liquid
Color Colorless
Density 0.9800 g/mL
Deuterium (D) 99.4% min.
Packaging Glass Bottle
Water 0.06% max. (Coulometric)
Quantity 25 mL
IUPAC Name (²H₈)oxolane
Formula Weight 80.17
Grade NMR
Chemical Name or Material Tetrahydrofuran-D8, 99.5 atom % D
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Tetrahydrofuran-D8
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Tetrahydrofuran

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.