missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofuran-d8, for NMR, packaged in 0.75 ml ampoules, 99.5 atom % D

Product Code. 10227430 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
7.5 mL
Packaging:
Ampule
Unit Size:
7.5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10227430 7.5 mL Ampule 7.5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10227430 Supplier Thermo Scientific Acros Supplier No. 320780075
  • ONLINE EXCLUSIVE PRICE
    334.00 €
    324.00 € / 7.5mL
    Save 10.00 €
    2% Off
View all promotions
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 1693-74-9 | C4H8O | 80.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Specifications

Physical Form Liquid
Color Colorless
Density 0.9800g/mL
Deuterium (D) 99.4% min.
Flash Point −21°C
Packaging Ampule
Refractive Index 1.407
Water 0.06% max. (Coulometric)
Quantity 7.5 mL
CAS 1693-74-9
Melting Point -106.0°C
Boiling Point 65.0°C to 66.0°C
Molecular Formula C4H8O
MDL Number MFCD00044238
Synonym tetrahydrofuran-d8, deuterated thf, octadeuterotetrahydrofuran, deuterated tetrahydrofuran, tetrahydrofuran-d8 thf-d8, tetrahydrofuran-d8, ≥99.5 atom % d, 2h4 tetrahydro 2h4 furan, thf-d8, 2h8 tetrahydrofuran, furan-d4-, tetrahydro-d4
InChI Key WYURNTSHIVDZCO-SVYQBANQSA-N
SMILES [2H]C1([2H])OC([2H])([2H])C([2H])([2H])C1([2H])[2H]
IUPAC Name 2,2,3,3,4,4,5,5-octadeuteriooxolane
Molecular Weight (g/mol) 80.16
PubChem CID 80290
Formula Weight 80.17
Grade NMR
Chemical Name or Material Tetrahydrofuran-d8, 99.5 atom % D
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Tetrahydrofuran-D8
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.