missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofuran, Extra Pure, SLR, Stabilised with 0.025% BHT, Fisher Chemical™

Product Code. 10152350 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
5 L
25 L
Packaging:
Amber Glass Bottle
HDPE Bottle
Metal Can
Plastic-coated Glass Bottle
Unit Size:
2.5 L
25 L
5 L
500mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10152350 25 L Metal Can 25 L
10559580 500 mL Amber Glass Bottle 500mL
10385860 2.5 L Amber Glass Bottle 2.5 L
10040380 2.5 L Plastic-coated Glass Bottle 2.5 L
10550831 5 L HDPE Bottle 5 L
10529960 5 L Metal Can 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 10152350 Supplier Fisher Chemical Supplier No. T/0700/25
  • ONLINE EXCLUSIVE PRICE
    695.00 €
    688.00 € / 25 L
    Save 7.00 €
    1% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

C4H8O, CAS Number-109-99-9, oxacyclopentane, tetraidrofurano, 1,4-epoxybutane, furan, tetrahydro, butylene oxide, hydrofuran, tetrahydrofuranne, tetrahydrofuran, furanidine, tetramethylene oxide, 2.5L, 66 deg.C, CHEBI:26911, Colorless, 72.11g/mol, WYURNTSHIVDZCO-UHFFFAOYSA-N, oxolane, -108.4 deg.C

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-99-9
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00005356
InChI Key WYURNTSHIVDZCO-UHFFFAOYSA-N
Synonym tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
PubChem CID 8028
ChEBI CHEBI:26911
IUPAC Name oxolane
SMILES C1CCOC1

Specifications

pH 7 to 8
Melting Point -108.4°C
Color Colorless
Quantity 25 L
Packaging Metal Can
Physical Form Liquid
Vapor Pressure 200mbar at 20°C
Viscosity 0.55cp at 20°C
Boiling Point 66°C
Formula Weight 72.11g/mol
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tetrahydrofuran
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.