missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrahydrofuran, Puriss. p.a., Reagent ACS, Stabilized, Honeywell Burdick & Jackson™
Click to view available options
:
200 L
Specifications
Specifications
| Melting Point | -108°C |
| CAS | 109-99-9 |
| Color | Colorless |
| Density | 0.89 g/cm3 (20°C) |
| Boiling Point | 65°C (1013 hPa) |
| Quantity | 200 L |
| Flash Point | -21°C |
| Molecular Formula | C4H8O |
| MDL Number | MFCD00005356 |
| Beilstein | 102391 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction