missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrahydrofuran (Sequencing), Fisher BioReagents
Description
Tetrahydrofuran is commonly used in sequencing procedures.
EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.
Specifications
Specifications
| pH | 7 to 8 |
| Melting Point | -108.4°C |
| Absorbance | 0.10 AU max. at 254nm, 0.30 AU max. at 230nm, 0.60 AU max. at 215nm, 1.00 AU max. at 210nm |
| CAS | 109-99-9 |
| Color | Colorless |
| Density | 0.880g/cm³ |
| Quantity | 1 L |
| Packaging | EcoSafPak™ Amber Glass |
| Physical Form | Liquid |
| Refractive Index | 1.4040 to 1.4070 (at 25°C) |
| Show More |
Safety and Handling
Product Identifier
- Tetrahydrofuran
- Danger
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H351-Suspected of causing cancer.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H302-Harmful if swallowed.
- H225-Highly flammable liquid and vapour.
- EUH019-May form explosive peroxides.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?