missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofuran, Spectrophotometric Grade, 99.7+%, unstab.

Product Code. 11388397
Change view
Click to view available options
Quantity:
100 mL
1 L
4 x 1 L
Unit Size:
1 L
100mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11388397 1 L 1 L
11318407 100 mL 100mL
11308407 1 L 1 L
15486065 4 x 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11388397 Supplier Thermo Scientific Alfa Aesar Supplier No. 032468.K2
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 109-99-9 | C4H8O | 72.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-99-9
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00005356
InChI Key WYURNTSHIVDZCO-UHFFFAOYSA-N
Synonym tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
PubChem CID 8028
ChEBI CHEBI:26911
IUPAC Name oxolane
SMILES C1CCOC1

Specifications

Melting Point °C to 108°C
Color Colorless
Density 0.889
Quantity 1 L
Flash Point -17°C (1°F)
Physical Form Liquid, Distilled In Glass
Refractive Index 1.407
Assay Percent Range >99.7%
Beilstein 102391
Boiling Point 66°C
Solubility Information Miscible with water.
Formula Weight 72.11
Grade Spectrophotometry
Chemical Name or Material Tetrahydrofuran, Filtered through 0.2 micron filters. Suitable for spectophotometry
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.