missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofurfuryl acrylate, tech. 90%, stab.

Product Code. 11308365
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11308365 50 g 50g
11318365 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11308365 Supplier Thermo Scientific Alfa Aesar Supplier No. L10299.18
This item is not returnable. View return policy

CAS: 2399-48-6 | C8H12O3 | 156.181 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2399-48-6
Molecular Formula C8H12O3
Molecular Weight (g/mol) 156.181
MDL Number MFCD00014516
InChI Key YNXCGLKMOXLBOD-UHFFFAOYSA-N
Synonym tetrahydrofurfuryl acrylate, acrylic acid, tetrahydrofurfuryl ester, 2-propenoic acid, tetrahydro-2-furanyl methyl ester, sartomer 285, sartomer 302, viscoat 150, kayarad tc 101, light acrylate thf-a, thf-a, 2-propenoic acid, tetrahydrofurfuryl ester
PubChem CID 94232
IUPAC Name oxolan-2-ylmethyl prop-2-enoate
SMILES C=CC(=O)OCC1CCCO1

Specifications

Density 1.03
Boiling Point 88°C to 89°C (9 mmHg)
Flash Point 93°C (199°F)
Refractive Index 1.455
Quantity 50 g
UN Number UN3334
Beilstein 113115
Formula Weight 156.18
Percent Purity 90%
Grade Technical
Chemical Name or Material Tetrahydrofurfuryl acrylate, stabilized with 500ppm hydroquinone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.