missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofurfuryl alcohol, 98%

Product Code. 10674992 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10674992 2.5 L Glass bottle 2.5 L
10235380 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10674992 Supplier Thermo Scientific Acros Supplier No. 138180025
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 97-99-4 | C5H10O2 | 102.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-99-4
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
MDL Number MFCD00005372
InChI Key BSYVTEYKTMYBMK-UHFFFAOYNA-N
Synonym tetrahydrofurfuryl alcohol, tetrahydro-2-furanmethanol, tetrahydrofuran-2-yl methanol, thfa, 2-furanmethanol, tetrahydro, tetrahydro-2-furanylmethanol, qo thfa, tetrahydro-2-furancarbinol, tetrahydrofuryl carbinol, furfuryl alcohol, tetrahydro
PubChem CID 7360
SMILES OCC1CCCO1

Specifications

Melting Point -80.0°C
Density 1.0543g/mL
Quantity 2.5 L
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.4510 to 1.4530
Assay Percent Range 97.5% min. (GC)
Specific Gravity 1.0543
Viscosity 6.24 mPa.s (20°C)
Beilstein 17, V,3, 115
Merck Index 15, 9358
Boiling Point 178.0°C
Solubility Information Solubility in water: soluble. Other solubilities: miscible with alcohol, ether, acetone, chloroform, and benzene
IUPAC Name oxolan-2-ylmethanol
Formula Weight 102.13
Percent Purity 98%
Chemical Name or Material Tetrahydrofurfuryl alcohol, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Tetrahydrofurfuryl alcohol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
Hazard Statement
  • H319-Causes serious eye irritation.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Tetrahydrofurfuryl alcohol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.