missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofurfuryl Alcohol, TRC

Product Code. 30451870
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30451870 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30451870 Supplier Toronto Research Chemicals Supplier No. TRCT79565310ML
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetrahydrofurfuryl alcohol
CAS 97-99-4
Molecular Formula C5 H10 O2
Quantity 10 mL
Synonym Furfuryl alcohol','tetrahydro- (8CI), Tetrahydro-2-furanmethanol, (Tetrahydrofuran-2-yl)methanol, (±)-Tetrahydrofurfuryl alcohol, 2-(Hydroxymethyl)tetrahydrofuran, NSC 15434, QO THFA, Rac-(Tetrahydrofuran-2-yl)methanol, THFA, Tetrahydro-2-furancarbinol, Tetrahydro-2-furanylmethanol, Tetrahydro-2-furylmethanol
InChI Formula InChI=1S/C5H10O2/c6-4-5-2-1-3-7-5/h5-6H,1-4H2
SMILES OCC1CCCO1
IUPAC Name oxolan-2-ylmethanol
Molecular Weight (g/mol) 102.1317
Formula Weight 102.0681
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.