missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofurfuryl bromide, 95%

Product Code. 11483940
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
Product Code. Quantity unitSize
11483940 5 g 5g
1 options
This item is not returnable. View return policy

Product Code. 11483940

Brand: Thermo Scientific Alfa Aesar A15192.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1192-30-9 | C5H9BrO | 165.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1192-30-9
Density 1.452
Quantity 5 g
Refractive Index 1.488
Molecular Formula C5H9BrO
MDL Number MFCD00016894
Beilstein 102717
Synonym tetrahydrofurfuryl bromide, 2-bromomethyl tetrahydrofuran, 2-bromomethyl oxolane, furan, 2-bromomethyl tetrahydro, 2-bromomethyl-tetrahydrofuran
InChI Key VOHILFSOWRNVJJ-UHFFFAOYSA-N
SMILES C1CC(OC1)CBr
IUPAC Name 2-(bromomethyl)oxolane
Molecular Weight (g/mol) 165.03
PubChem CID 14501
Formula Weight 165.04
Percent Purity 95%
Chemical Name or Material Tetrahydrofurfuryl bromide
Show More Show Less
compliance-icons
Product Identifier
  • Tetrahydrofurfuryl bromide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Tetrahydrofurfuryl bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.