missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydronaphthalene, TRC

Product Code. 30424530
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
Product Code. Quantity unitSize
30424530 25 g 25g
1 options
This item is not returnable. View return policy

Product Code. 30424530

Brand: Toronto Research Chemicals TRCT29399625G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,2,3,4-Tetrahydronaphthalene
CAS 119-64-2
Molecular Formula C10 H12
Quantity 25 g
Synonym 1,2,3,4-Tetrahydronaphthalene, Naphthalene','1,2,3,4-tetrahydro-, 1,2,3,4-Tetrahydronaphthalene, Benzocyclohexane, NSC 77451, Tetrahydronaphthalene, Tetralin, Tetranap
InChI Formula InChI=1S/C10H12/c1-2-6-10-8-4-3-7-9(10)5-1/h1-2,5-6H,3-4,7-8H2
SMILES C1CCc2ccccc2C1
IUPAC Name 1,2,3,4-tetrahydronaphthalene
Molecular Weight (g/mol) 132.2
Formula Weight 132.0939
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.