missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydronaphthalene, TRC

Product Code. 30422334
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30422334 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30422334 Supplier Toronto Research Chemicals Supplier No. TRCT29399610G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,2,3,4-Tetrahydronaphthalene
CAS 119-64-2
Molecular Formula C10 H12
Quantity 10 g
Synonym 1,2,3,4-Tetrahydronaphthalene, Naphthalene','1,2,3,4-tetrahydro-, 1,2,3,4-Tetrahydronaphthalene, Benzocyclohexane, NSC 77451, Tetrahydronaphthalene, Tetralin, Tetranap
InChI Formula InChI=1S/C10H12/c1-2-6-10-8-4-3-7-9(10)5-1/h1-2,5-6H,3-4,7-8H2
SMILES C1CCc2ccccc2C1
IUPAC Name 1,2,3,4-tetrahydronaphthalene
Molecular Weight (g/mol) 132.2
Formula Weight 132.0939
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.