missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydropalmatine, MedChemExpress

Product Code. 16494807
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16494807 50 mg 50mg
16408948 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16494807 Supplier Medchem Express Supplier No. HYN0300
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Tetrahydropalmatine possesses analgesic effects. Tetrahydropalmatine acts through inhibition of amygdaloid release of dopamine to inhibit an epileptic attack in rats.

Tetrahydropalmatine possesses analgesic effects. Tetrahydropalmatine acts through inhibition of amygdaloid release of dopamine to inhibit an epileptic attack in rats.

Specifications

Chemical Name or Material Tetrahydropalmatine
CAS 2934-97-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C21H25NO4
Quantity 50 mg
Synonym DL-Tetrahydropalmatine
Solubility Information DMSO : 6.67 mg/mL (18.77 mM; Need ultrasonic)
SMILES COC1=CC=C2C(CN3CCC4=CC(OC)=C(OC)C=C4C3C2)=C1OC
Molecular Weight (g/mol) 355.43
Formula Weight 355.43
Percent Purity 99.07%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.