missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydropyran-2-methanol, 98%, Thermo Scientific Chemicals

Product Code. 10681253 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10681253 10 g Glass bottle 10g
10386970 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10681253 Supplier Thermo Scientific Acros Supplier No. 138250100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 100-72-1 | Molecular Formula: C6H12O2 | Molecular Weight: 116.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-72-1
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00006624
InChI Key ROTONRWJLXYJBD-UHFFFAOYSA-N
Synonym tetrahydropyran-2-methanol, 2h-pyran-2-methanol, tetrahydro, 2-hydroxymethyltetrahydropyran, tetrahydropyran-2-carbinol, 2-hydroxymethyl tetrahydropyran, pyran-2-methanol, tetrahydro, tetrahydro-2h-pyran-2-yl methanol, 2-tetrahydropyranilcarbinol, tetrahydropyranyl-2-methanol, 2-methyloltetrahydro-1,4-pyran
PubChem CID 7524
IUPAC Name oxan-2-ylmethanol
SMILES C1CCOC(C1)CO

Specifications

Density 1.0200g/mL
Boiling Point 187°C
Flash Point 93°C
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.4570 to 1.459
Quantity 10 g
Specific Gravity 1.02
Formula Weight 116.16
Percent Purity 98%
Chemical Name or Material Tetrahydropyran-2-methanol, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Tetrahydropyran-2-methanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.