missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydropyran-4-carbothioamide, 90%, Thermo Scientific™

Product Code. 10103034
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10103034 250 mg Amber Glass Bottle 250mg
10578934 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10103034 Supplier Thermo Scientific Maybridge Supplier No. CC29918CB
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 88571-77-1
Molecular Formula C6H11NOS
Molecular Weight (g/mol) 145.22
MDL Number MFCD10700048
InChI Key ZUONFEFOQCUTDW-UHFFFAOYSA-N
Synonym tetrahydro-2h-pyran-4-carbothioamide, tetrahydropyran-4-carbothioamide, 2h-pyran-4-carbothioamide,tetrahydro, 2h-pyran-4-carbothioamide, tetrahydro, acmc-20fauh, oxane-4-carboimidothioic acid, 4-carbamothioyl tetrahydro-2h-pyran, tetrahydro-2h-pyran-4-carbothioic acid amide, 2h-3,4,5,6-tetrahydropyran-4-ylaminomethane-1-thione
PubChem CID 13197204
IUPAC Name oxane-4-carbothioamide
SMILES C1COCCC1C(=S)N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 145.22
Percent Purity 97%
Chemical Name or Material Tetrahydropyran-4-carbothioamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.