missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydropyran-4-carboxylic acid, 97%

Product Code. 15375078 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15375078 25 g 25g
15365078 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15375078 Supplier Thermo Scientific Acros Supplier No. 456720250
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5337-03-1 | C6H10O3 | 130.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5337-03-1
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.14
InChI Key AVPKHOTUOHDTLW-UHFFFAOYSA-N
Synonym tetrahydro-2h-pyran-4-carboxylic acid, tetrahydropyran-4-yl-carboxylic acid, tetrahydropyran-4-carboxylic acid, tetrahydro-pyran-4-carboxylic acid, 2h-pyran-4-carboxylic acid, tetrahydro, 4-carboxytetrahydro-2h-pyran, 3,4,5,6-tetrahydro-2h-pyran-4-carboxylic acid, 2h-3,4,5,6-tetrahydropyran-4-carboxylic acid, pubchem9730, acmc-2097ha
PubChem CID 219302
IUPAC Name oxane-4-carboxylic acid
SMILES C1COCCC1C(=O)O

Specifications

Melting Point 87.0°C to 89.0°C
Boiling Point 115.0°C (20.0 mmHg)
Infrared Spectrum Authentic
Quantity 25 g
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol
Formula Weight 130.14
Percent Purity 97%
Chemical Name or Material Tetrahydropyran-4-carboxylic acid
compliance-icons
Product Identifier
  • Tetrahydropyran-4-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.