missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydropyran, TRC

Product Code. 30412791
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30412791 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30412791 Supplier Toronto Research Chemicals Supplier No. TRCT29503025G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetrahydropyran
CAS 142-68-7
Molecular Formula C5 H10 O
Quantity 25 g
Synonym 2H-Pyran','tetrahydro-, Pyran','tetrahydro- (3CI), Tetrahydro-2H-pyran, NSC 65448, Oxacyclohexane, Oxane, Pentamethylene oxide, THP, Tetrahydropyran, Tetrahydropyrane
InChI Formula InChI=1S/C5H10O/c1-2-4-6-5-3-1/h1-5H2
SMILES C1CCOCC1
IUPAC Name oxane
Molecular Weight (g/mol) 86.132
Formula Weight 86.0732
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.