missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrothiophene 1-oxide, 97%

Product Code. 11418117
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11418117 50 g 50g
11408117 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11418117 Supplier Thermo Scientific Alfa Aesar Supplier No. A17502.18
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 1600-44-8 | C4H8OS | 104.167 g/mol

In a comparative study of aromatic fluorodenitration with Potassium fluoride, in several dipolar aprotic solvents, a much faster reaction rate was observed with DMSO or tetramethylene sulfoxide than with the more commonly used Sulfolane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
In a comparative study of aromatic fluorodenitration with Potassium fluoride, in several dipolar aprotic solvents, a much faster reaction rate was observed with DMSO or tetramethylene sulfoxide than with the more commonly used Sulfolane.

Solubility
Fully miscible in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 1600-44-8
Molecular Formula C4H8OS
Molecular Weight (g/mol) 104.167
MDL Number MFCD00005477
InChI Key ISXOBTBCNRIIQO-UHFFFAOYSA-N
Synonym tetramethylene sulfoxide, tetrahydrothiophene 1-oxide, thiophane oxide, tetrahydrothiophene oxide, thiophane monoxide, thiophane 1-oxide, thiophene, tetrahydro-, 1-oxide, tetramethylene sulphoxide, thiophene, tetrahydro, 1-oxide, tetrametylene sulfoxide
PubChem CID 1128
IUPAC Name thiolane 1-oxide
SMILES C1CCS(=O)C1

Spécification

Density 1.173
Boiling Point 235°C to 237°C
Flash Point 112°C (233°F)
Refractive Index 1.52
Quantity 50 g
Beilstein 105274
Solubility Information Fully miscible in water.
Formula Weight 104.17
Percent Purity 97%
Chemical Name or Material Tetrahydrothiophene 1-oxide

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.