missing translation for 'onlineSavingsMsg'
Learn More

tetrahydrothiopyran-4-carbonitrile, 97%, Thermo Scientific™

Product Code. 10442684
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10442684 5 g Amber Glass Bottle 5g
10012514 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10442684 Supplier Thermo Scientific Maybridge Supplier No. CC61916DE
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 195503-40-3
Molecular Formula C6H9NS
Molecular Weight (g/mol) 127.21
MDL Number MFCD09025909
InChI Key DQKVZWAEWJUKAX-UHFFFAOYSA-N
Synonym tetrahydro-2h-thiopyran-4-carbonitrile, tetrahydrothiopyran-4-carbonitrile, 2h-thiopyran-4-carbonitrile,tetrahydro, 2h-thiopyran-4-carbonitrile, tetrahydro, 4-cyanothiane, 4-cyanothian-4-yl, tetrahydro-thiopyran-4-carbonitrile
PubChem CID 21598641
IUPAC Name thiane-4-carbonitrile
SMILES N#CC1CCSCC1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 127.21
Percent Purity 97%
Chemical Name or Material Tetrahydrothiopyran-4-carbonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.