missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrothiopyran-4-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10452834
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10452834 250 mg Amber Glass Bottle 250mg
10665974 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10452834 Supplier Thermo Scientific Maybridge Supplier No. CC61901CB
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 89489-53-2
Molecular Formula C6H10O2S
Molecular Weight (g/mol) 146.20
MDL Number MFCD09879921
InChI Key SCKSQOIXSGPVJS-UHFFFAOYSA-N
Synonym tetrahydro-2h-thiopyran-4-carboxylic acid, tetrahydrothiopyran-4-carboxylic acid, 2h-thiopyran-4-carboxylic acid, tetrahydro, 2h-thiopyran-4-carboxylicacid, tetrahydro, 2h-thiopyran-4-carboxylicacid,tetrahydro-9ci, acmc-209vjr, tetrahydro-thiopyran-4-carboxylic acid, 2h-thiopyran-4-carboxylic acid, tetrahydro-9ci
PubChem CID 21598642
IUPAC Name thiane-4-carboxylic acid
SMILES OC(=O)C1CCSCC1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 146.21
Percent Purity 97%
Chemical Name or Material Tetrahydrothiopyran-4-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.