missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrothiopyran-4-ylmethylamine hydrochloride, 97%, Thermo Scientific™

Product Code. 10403044
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10403044 250 mg Amber Glass Bottle 250mg
10534974 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10403044 Supplier Thermo Scientific Maybridge Supplier No. CC61963CB
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 950603-21-1
Molecular Formula C6H14ClNS
Molecular Weight (g/mol) 167.695
MDL Number MFCD09879925
InChI Key DREBXEFIPXBLAP-UHFFFAOYSA-N
Synonym tetrahydro-2h-thiopyran-4-yl methanamine hydrochloride, tetrahydrothiopyran-4-ylmethyl amine hydrochloride, thian-4-ylmethanamine hydrochloride, thian-4-ylmethylamine, chloride, 1-thian-4-yl methanamine hydrochloride, tetrahydrothiopyran-4-ylmethylamine hydrochloride, 1-thian-4-yl methanamine-hydrogen chloride 1/1, tetrahydro-2h-thiopyran-4-yl methylamine hydrochloride
PubChem CID 24229644
IUPAC Name thian-4-ylmethanamine;hydrochloride
SMILES C1CSCCC1CN.Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 167.7
Percent Purity 97%
Chemical Name or Material Tetrahydrothiopyran-4-ylmethylamine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.