missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrothiopyran, 98%

Product Code. 11312016
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11312016 1 g 1g
11322016 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 11312016

Brand: Thermo Scientific Alfa Aesar L03063.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1613-51-0 | C5H10S | 102.195 g/mol

It is a useful 5 c synthon for the synthesis of 3-cyclopentenones. It is a organic sulfide, that has shown to have antioxidant activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a useful 5 c synthon for the synthesis of 3-cyclopentenones. It is a organic sulfide, that has shown to have antioxidant activity.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources. Protect against electrostatic charges.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1613-51-0
Molecular Formula C5H10S
Molecular Weight (g/mol) 102.195
MDL Number MFCD00006662
InChI Key YPWFISCTZQNZAU-UHFFFAOYSA-N
Synonym pentamethylene sulfide, tetrahydrothiopyran, tetrahydro-2h-thiopyran, thiacyclohexane, 2h-thiopyran, tetrahydro, penthiophane, tetrahydrothiapyran, tetrahydrothiopyrane, thian, thian-4-yl
PubChem CID 15367
IUPAC Name thiane
SMILES C1CCSCC1

Specifications

Density 0.988
Boiling Point 140°C to 142°C
Flash Point 21°C (69°F)
Odor Unpleasant
Refractive Index 1.506
Quantity 1 g
UN Number UN1993
Beilstein 102437
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 102.2
Percent Purity 98%
Chemical Name or Material Tetrahydrothiopyran
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.