missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrothiopyran, 98%

Product Code. 11322016
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11322016 5 g 5g
11312016 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11322016 Supplier Thermo Scientific Alfa Aesar Supplier No. L03063.06
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1613-51-0 | C5H10S | 102.195 g/mol

It is a useful 5 c synthon for the synthesis of 3-cyclopentenones. It is a organic sulfide, that has shown to have antioxidant activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a useful 5 c synthon for the synthesis of 3-cyclopentenones. It is a organic sulfide, that has shown to have antioxidant activity.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources. Protect against electrostatic charges.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1613-51-0
Molecular Formula C5H10S
Molecular Weight (g/mol) 102.195
MDL Number MFCD00006662
InChI Key YPWFISCTZQNZAU-UHFFFAOYSA-N
Synonym pentamethylene sulfide, tetrahydrothiopyran, tetrahydro-2h-thiopyran, thiacyclohexane, 2h-thiopyran, tetrahydro, penthiophane, tetrahydrothiapyran, tetrahydrothiopyrane, thian, thian-4-yl
PubChem CID 15367
IUPAC Name thiane
SMILES C1CCSCC1

Specifications

Density 0.988
Boiling Point 140°C to 142°C
Flash Point 21°C (69°F)
Odor Unpleasant
Refractive Index 1.506
Quantity 5 g
UN Number UN1993
Beilstein 102437
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 102.2
Percent Purity 98%
Chemical Name or Material Tetrahydrothiopyran
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.