missing translation for 'onlineSavingsMsg'
Learn More

Tetraisopropyl methylenediphosphonate, 98%

Product Code. 11495563
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11495563 50 g 50g
11485563 10 g 10g
11405573 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11495563 Supplier Thermo Scientific Alfa Aesar Supplier No. A13484.18
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1660-95-3 | C13H30O6P2 | 344.325 g/mol

Tetraisopropyl methylenediphosphonate is used in the preparation of AZT 5’-Triphosphate compounds which display inhibitory effects on HIV-1 reverse transcrpitase. It is also used in the preparation of E2-bisphosphonate conjugates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetraisopropyl methylenediphosphonate is used in the preparation of AZT 5’-Triphosphate compounds which display inhibitory effects on HIV-1 reverse transcrpitase. It is also used in the preparation of E2-bisphosphonate conjugates.

Solubility
Insoluble in water.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1660-95-3
Molecular Formula C13H30O6P2
Molecular Weight (g/mol) 344.325
MDL Number MFCD00015021
InChI Key ODTQUKVFOLFLIQ-UHFFFAOYSA-N
Synonym tetraisopropyl methylenediphosphonate, phosphonic acid, methylenebis-, tetrakis 1-methylethyl ester, tetraisopropyl methylenebisphosphonate, unii-k46y5v9u61, tetraisopropyl methylenebis phosphonate, methylenediphosphonic acid tetraisopropyl ester, tetra-i-propylmethylenediphosphonate, 2-di propan-2-yloxy phosphorylmethyl-propan-2-yloxyphosphoryl oxypropane, phosphonic acid, p,p'-methylenebis-, p,p,p',p'-tetrakis 1-methylethyl ester, dsstox_cid_24847
PubChem CID 74266
IUPAC Name 2-[di(propan-2-yloxy)phosphorylmethyl-propan-2-yloxyphosphoryl]oxypropane
SMILES CC(C)OP(=O)(CP(=O)(OC(C)C)OC(C)C)OC(C)C

Specifications

Density 1.08
Boiling Point 122°C to 124°C (1.5 mmHg)
Flash Point 160°C (320°F)
Refractive Index 1.434
Quantity 50 g
Beilstein 1080180
Solubility Information Insoluble in water.
Formula Weight 344.33
Percent Purity 98%
Chemical Name or Material Tetraisopropyl methylenediphosphonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.