missing translation for 'onlineSavingsMsg'
Learn More

Tetrakis(acetonitrile)copper(I) tetrafluoroborate

Product Code. 10192414 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10192414 5 g Glass bottle 5g
10480894 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10192414 Supplier Thermo Scientific Acros Supplier No. 398760050
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 15418-29-8 | C8H12BCuF4N4 | 314.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15418-29-8
Molecular Formula C8H12BCuF4N4
Molecular Weight (g/mol) 314.56
MDL Number MFCD09265110
InChI Key YZGSKMIIVMCEFE-UHFFFAOYSA-N
PubChem CID 10990673
IUPAC Name acetonitrile;copper(1+);tetrafluoroborate
SMILES [Cu+].CC#N.CC#N.CC#N.CC#N.F[B-](F)(F)F

Specifications

Packaging Glass bottle
Quantity 5 g
Synonym tetrakis acetonitrile copper i tetrafluoroborate, cu mecn 4 bf4, cu mecn 4bf4, copper i tetra acetonitrile tetrafluoroborate, tetra acetonitrile copper 1+ tetrafluoroborate
Formula Weight 314.56
Chemical Name or Material Tetrakis(acetonitrile)copper(I) tetrafluoroborate
compliance-icons
Product Identifier
  • Tetrakis(acetonitrile)copper(I) tetrafluoroborate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.