missing translation for 'onlineSavingsMsg'
Learn More

Tetrakis(acetonitrile)palladium(II) Bis(trifluoromethanesulfonate) >90%, TRC

Product Code. 30422326
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30422326 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30422326 Supplier Toronto Research Chemicals Supplier No. TRCT296335100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetrakis(acetonitrile)palladium(II) Bis(trifluoromethanesulfonate)
CAS 68569-14-2
Molecular Formula C8 H12 N4 Pd . C F3 O3 S
Quantity 100 mg
Synonym Tetrakis(acetonitrile)palladium Bis(triflate), Tetrakis(acetonitrile)palladium Ditriflate, Tetrakis(acetonitrile)palladium(2+) Ditriflate, (SP-4-1)-Tetrakis(acetonitrile)palladium(2+) Salt With Trifluoromethanesulfonic Acid
InChI Formula InChI=1S/4C2H3N.CHF3O3S.Pd/c4*1-2-3;2-1(3,4)8(5,6)7;/h4*1H3;(H,5,6,7);/q;;;;;+2/p-1
SMILES CC#[N][Pd+2]([N]#CC)([N]#CC)[N]#CC.[O-]S(=O)(=O)C(F)(F)F
IUPAC Name tetrakis(ethylidyne-λ5-azanyl)palladium(2+);trifluoromethanesulfonate
Molecular Weight (g/mol) 419.697
Formula Weight 418.961
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.