missing translation for 'onlineSavingsMsg'
Learn More

Tetrakis(acetonitrile)palladium(II) tetrafluoroborate, 99%

Product Code. 10643763 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10643763 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10643763 Supplier Thermo Scientific Acros Supplier No. 363520010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 21797-13-7 | C8H12B2F8N4Pd | 444.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 21797-13-7
Melting Point 230°C
Color Yellow
Elemental Analysis 98.5% min.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C8H12B2F8N4Pd
MDL Number MFCD00043297
Quantity 1 g
Synonym tetrakis acetonitrile palladium ii tetrafluoroborate, acetonitrile; palladium 2+ ; ditetrafluoroborate, pd mecn 4 bf4 2, tetrakis acetonitrile palladium ii, palladium ii tetrafluoroborate tetraacetonitrile complex
Solubility Information Solubility in water: soluble in water
InChI Key YWMRPVUMBTVUEX-UHFFFAOYSA-N
SMILES [Pd++].CC#N.CC#N.CC#N.CC#N.F[B-](F)(F)F.F[B-](F)(F)F
IUPAC Name acetonitrile;palladium(2+);ditetrafluoroborate
Molecular Weight (g/mol) 444.24
PubChem CID 2734560
Formula Weight 444.22
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Tetrakis(acetonitrile)palladium(II) tetrafluoroborate
Show More Show Less
compliance-icons
Product Identifier
  • Tetrakis(acetonitrile)palladium(II) tetrafluoroborate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.