missing translation for 'onlineSavingsMsg'
Learn More

Tetrakis(dimethylamino)zirconium

Product Code. 10402683 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10402683 1 g Glass bottle 1g
10538473 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10402683 Supplier Thermo Scientific Acros Supplier No. 380410010
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 19756-04-8 | C8H24N4Zr | 267.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19756-04-8
Molecular Formula C8H24N4Zr
Molecular Weight (g/mol) 267.53
MDL Number MFCD00239502
InChI Key DWCMDRNGBIZOQL-UHFFFAOYSA-N
Synonym tetrakis dimethylamino zirconium, zirconium, tetrakis dimethylamino, tdmaz, tetrakis dimethylamino zirconium iv, tetrakis dimethylamido zirconium iv, zr nme2 4, dimethyl tris dimethylamino zirconio amine, tetrakis dimethylamido zirconium iv , crystalline, tetrakis dimethylamido zirconium iv , deposition grade, tetrakis dimethylamido zirconium iv , packaged for use in deposition systems
PubChem CID 140580
IUPAC Name dimethylazanide;zirconium(4+)
SMILES C[N-]C.C[N-]C.C[N-]C.C[N-]C.[Zr+4]

Specifications

Melting Point 57°C to 60°C
Color Green-Yellow
Packaging Glass bottle
Quantity 1 g
Formula Weight 267.53
Physical Form Crystalline Solid
Chemical Name or Material Tetrakis(dimethylamino)zirconium

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.