missing translation for 'onlineSavingsMsg'
Learn More

Tetramethrin, TRC

Product Code. 30415105
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30415105 10 g 10g
30409317 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30415105 Supplier Toronto Research Chemicals Supplier No. TRCT29869010G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7696-12-0
Molecular Formula C19 H25 N O4
Molecular Weight (g/mol) 331.41
Synonym Tetramethrin, Phthalthrin, Cyclopropanecarboxylic acid, 2,2-dimethyl-3-(2-methyl-1-propen-1-yl)-, (1,3,4,5,6,7-hexahydro-1,3-dioxo-2H-isoindol-2-yl)methyl ester (ACI), Cyclopropanecarboxylic acid, 2,2-dimethyl-3-(2-methyl-1-propenyl)-, (1,3,4,5,6,7-hexahydro-1,3-dioxo-2H-isoindol-2-yl)methyl ester (9CI), Cyclopropanecarboxylic acid, 2,2-dimethyl-3-(2-methylpropenyl)-, ester with N-(hydroxymethyl)-1-cyclohexene-1,2-dicarboximide (7CI, 8CI), (1,3-Dioxo-4,5,6,7-tetrahydro-1H-isoindol-2(3H)-yl)methyl 2,2-dimethyl-3-(2-methylprop-1-en-1-yl)cyclopropanecarboxylate, (1-Cyclohexene-1,2-dicarboximido)methyl chrysanthemate, (1-Cyclohexene-1,2-dicarboximido)methyl chrysanthemumate, Bioneo-Pynamin, d-Tetramethrin, Insectol, Neo-Pynamin, Neo-Pynamin Forte, Phthalthrin, Pibutox, Pion 1, Pyrethrol L, Tetramethrin
IUPAC Name (1,3-dioxo-4,5,6,7-tetrahydroisoindol-2-yl)methyl 2,2-dimethyl-3-(2-methylprop-1-enyl)cyclopropane-1-carboxylate
SMILES CC(=CC1C(C(=O)OCN2C(=O)C3=C(CCCC3)C2=O)C1(C)C)C

Specifications

Quantity 10 g
InChI Formula InChI=1S/C19H25NO4/c1-11(2)9-14-15(19(14,3)4)18(23)24-10-20-16(21)12-7-5-6-8-13(12)17(20)22/h9,14-15H,5-8,10H2,1-4H3
Formula Weight 331.1784
Chemical Name or Material Tetramethrin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.