missing translation for 'onlineSavingsMsg'
Learn More

Tetramethyl Ammonium Hydroxide, Extra Pure, Solution 25% In Water, SLR, Fisher Chemical™

Product Code. 10558260 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10558260 25 mL 25mL
10132450 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Código de producto. 10558260 Proveedor Fisher Chemical N.º de proveedor T/0940/04
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos

Este artículo no se puede devolver. Vea la política de devoluciones

Identificadores de productos químicos

CAS 75-59-2
Molecular Formula C4H13NO
Molecular Weight (g/mol) 91.154
MDL Number 8280
InChI Key WGTYBPLFGIVFAS-UHFFFAOYSA-M
Synonym tetramethylammonium hydroxide, tmah, hydroxyde de tetramethylammonium, nmw-w, nmd 3, tetramethyl ammonium hydroxide, unii-5gkp7317q2, ammonium, tetramethyl-, hydroxide, methanaminium, n,n,n-trimethyl-, hydroxide, tetramethylammoniumhydroxide
PubChem CID 60966
IUPAC Name tetramethylazanium;hydroxide
SMILES C[N+](C)(C)C.[OH-]

Especificaciones

Boiling Point 102°C
Melting Point -25°C
Color Yellow
pH 13
Physical Form Liquid
Quantity 25 mL
Formula Weight 91.15g/mol
Packaging HDPE plastic bottle
Vapor Pressure 16mmHg (25°C)
compliance-icons compliance-icons compliance-icons compliance-icons
Nombre del producto
  • Tetramethylammonium hydroxide, 25% in water
Palabra de advertencia
  • Danger
Clasificación
  • Acute toxicity Category 3
  • Acute toxicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity after single exposure Category 1
  • Specific target organ toxicity Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
Indicaciones de peligro
  • H301-Toxic if swallowed.
  • H310-Fatal in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H370-Causes damage to organs.
  • H372-Causes damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
Consejos de prudencia
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
Otros peligros
  • MIXTURE LIST-Contain: Tetramethylammoniumhydroxide
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.