missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium fluoride tetrahydrate, 98%

Product Code. 10041660 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10041660 5 g Glass bottle 5g
10397070 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10041660 Supplier Thermo Scientific Acros Supplier No. 148070050
  • ONLINE EXCLUSIVE PRICE
    35.65 €
    33.40 € / 5g
    Save 2.25 €
    6% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 17787-40-5 | C4H12FN·4H2O | 165.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17787-40-5
Molecular Formula C4H12FN·4H2O
Molecular Weight (g/mol) 165.21
MDL Number MFCD00149968
InChI Key HQFTZNVQVRRDLN-UHFFFAOYSA-M
Synonym tetramethylammonium fluoride tetrahydrate, tetramethylazanium fluoride tetrahydrate, methanaminium, n,n,n-trimethyl-, fluoride, tetrahydrate, tetramethylammonium tetrahydrate fluoride, tetramethylammonium ion tetrahydrate fluoride, acmc-1c8r1, tetramethylammoniumfluoridetetrahydrate, methanaminium,n,n,n-trimethyl-,fluoride,tetrahydrate 9ci, tetramethylamine, fluoride, hydrate, hydrate, hydrate, hydrate, methanaminium,n,n,n-trimethyl-, fluoride, tetrahydrate 9ci
PubChem CID 2723808
IUPAC Name tetramethylazanium;fluoride;tetrahydrate
SMILES C[N+](C)(C)C.O.O.O.O.[F-]

Specifications

Melting Point 39°C to 44°C
Flash Point >112°C
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (CH3)4NF·4H2O
Quantity 5 g
Fieser 07,358
Solubility Information Solubility in water: soluble
Formula Weight 165.21
Percent Purity 98%
Chemical Name or Material Tetramethylammonium fluoride tetrahydrate, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Tetramethylammonium fluoride tetrahydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.